Learn More
ATCC HCC1395 BL; B Lymphoblast; Genomic DNA, Human
Catalog No. 502383481
Encompass
High molecular weight genomic DNA isolated from the HCC1395 BL EBV-transformed lymphoblastoid cell line (ATCC CRL-2325).
Catalog No. 50-238-3481 Supplier ATCC Supplier No. CRL2325D
High molecular weight genomic DNA isolated from the HCC1395 BL EBV-transformed lymphoblastoid cell line (ATCC CRL-2325).