Learn More
ATCC HCC1419; Breast Ductal Carcinoma; Human (Homo sapiens)
Catalog No. 502382119
Encompass
HCC1419 is an epithelial cell that was isolated from the mammary gland of a 42-year-old, Hispanic female patient with dutcal carcinoma TNM stage IIIA, grade 3. The cell was deposited by AF Gazdar and AK Virmani and can be used in cancer research.
Catalog No. 50-238-2119 Supplier ATCC Supplier No. CRL2326
HCC1419 is an epithelial cell that was isolated from the mammary gland of a 42-year-old, Hispanic female patient with dutcal carcinoma TNM stage IIIA, grade 3. The cell was deposited by AF Gazdar and AK Virmani and can be used in cancer research.