Learn More
ATCC HCC1500; Breast Ductal Carcinoma; Human (Homo sapiens)
Catalog No. 502383212
Encompass
HCC1500 is an epithelial cell that was isolated from the mammary gland of a 32-year-old, Black female patient with ductal carcinoma TNM stage IIB, grade 2. The cell was deposited by AF Gazdar and AK Virmani and can be used in cancer research.
Catalog No. 50-238-3212 Supplier ATCC Supplier No. CRL2329
HCC1500 is an epithelial cell that was isolated from the mammary gland of a 32-year-old, Black female patient with ductal carcinoma TNM stage IIB, grade 2. The cell was deposited by AF Gazdar and AK Virmani and can be used in cancer research.