Learn More
ATCC HCC1569; Breast Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL2330
Encompass
HCC1569 is an epithelial cell line isolated from the mammary gland of a 70-year-old, Black, female, metaplastic carcinoma patient with a germline mutation in the FHIT gene, which her daughter carries. This cell line was initiated on in 1995 and took 19 months to establish.
Catalog No. 50-238-3472