Learn More
ATCC HCC1599; Breast Carcinoma; Human
Catalog No. 502385085
Encompass
HCC1599 is an epithelial cell that was isolated from the mammary gland of a 44-year-old, White, female patient with ductal carcinoma TNM stage IIIA, grade 3. The cell was deposited by AF Gazdar and AK Virmani and can be used in cancer research.
Catalog No. 50-238-5085 Supplier ATCC Supplier No. CRL2331
HCC1599 is an epithelial cell that was isolated from the mammary gland of a 44-year-old, White, female patient with ductal carcinoma TNM stage IIIA, grade 3. The cell was deposited by AF Gazdar and AK Virmani and can be used in cancer research.