Learn More
ATCC HCC1806; Breast Carcinoma; Human (Homo sapiens)
Catalog No. 502383086
Encompass
HCC1806 is an epithelial cell line isolated from the mammary gland of a 60-year-old, Black, female patient with acantholytic squamous cell carcinoma (ASCC), TNM Stage IIB, grade 2. The HCC1806 breast cancer cell line was initiated in 1995 and took 10 months to establish.
Catalog No. 50-238-3086 Supplier ATCC Supplier No. CRL2335
Add to Cart
Add to Cart
HCC1806 is an epithelial cell line isolated from the mammary gland of a 60-year-old, Black, female patient with acantholytic squamous cell carcinoma (ASCC), TNM Stage IIB, grade 2. The HCC1806 breast cancer cell line was initiated in 1995 and took 10 months to establish.