Learn More
ATCC HCC2218 BL; Lymphoblast; Human (Homo sapiens)
Catalog No. 502384145
Encompass
HCC2218 BLis a normal B lymphoblast cell that was isolated from the peripheral blood of a 38-year-old, White, female patient with breast ductal carcinoma. The cell can be used in cancer and immunology research.
Catalog No. 50-238-4145 Supplier ATCC Supplier No. CRL2363
HCC2218 BLis a normal B lymphoblast cell that was isolated from the peripheral blood of a 38-year-old, White, female patient with breast ductal carcinoma. The cell can be used in cancer and immunology research.