Learn More
ATCC HCC70; Breast Carcinoma; Human (Homo sapiens)
Catalog No. 502384936
Encompass
HCC70 is an epithelial cell line isolated from a primary ductal carcinoma from a 49-year-old, Black female in 1992, and the cells took 44 months to establish. HCC70 is positive for the epithelial cell specific marker Epithelial Glycoprotein 2 (EGP2) and for cytokeratin 19.
Catalog No. 50-238-4936 Supplier ATCC Supplier No. CRL2315
HCC70 is an epithelial cell line isolated from a primary ductal carcinoma from a 49-year-old, Black female in 1992, and the cells took 44 months to establish. HCC70 is positive for the epithelial cell specific marker Epithelial Glycoprotein 2 (EGP2) and for cytokeratin 19.