Learn More
Aviva Systems Biology Corp HCCS Antibody - N-terminal region (ARP61091_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61091_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HCCS. It was validated on Western Blot. . Alternate names: CCHL, DKFZp779I1858, MCOPS7ApplicationWB. Species reactivity: Dog, Horse, Human, Rat
Catalog No. 50-287-248