Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hourglass International™ HCE™ 2-Ply Cleanroom Face Mask with Ties
BIOPHARMA

Catalog No. 180401270 Shop All Hourglass International Inc Products
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-040-1270 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-040-1270 Supplier Hourglass International™ Supplier No. FM2ESB23221
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cleanroom face mask with four white ties and a soft flexible nose piece are made from the cleanest polyethylene and polypropylene materials. Suitable for ISO 5 and higher cleanroom environments.

  • Ultra-low lint non-woven ES and SBPP materials
  • Four white SBPP ties for a custom fit
  • Soft, flexible nose piece
  • Ties are attached to the mask body for strength
  • Manufactured and double-bagged in a cleanroom enviroment for particle control
  • Statistical manufacturing process controls ensure consistency and quality lot after lot
  • Suitable for ISO 5/Class 100 cleanroom environments and up

Specifications

Product Type Cleanroom Face Mask
Color White
Fastening Type Ties
Material Polypropylene
Size Large
Sterility Non-sterile
Style Pleated
Certifications/Compliance ISO 9001
Cleanroom Class Class 100 (ISO 5)
Construction 2-ply
Disposable Single-use
For Use With (Application) Semiconductor, Disk Drive, Medical Device, Pharmaceutical, Biomedical
Latex Free Yes
Length (English) 8 in.
Length (Metric) 21 cm
Model FM2ESB232-21
Packaging Quantity 50/Bag; 2000/Cs.
Packaging Type Double-bagged, Cleanroom packaged
Series HCE™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.