Learn More
Promocell GmbH PromoCell HCF-c 500,000 Human Cardiac Fibroblasts, cryopreserved
Catalog No. 50306008
Encompass
Primary Human Cardiac Fibroblasts (HCF) are isolated from the ventricles of the adult heart. They play a central role in the maintenance of the extracellular matrix in the normal heart and the synthesis of growth factors and cytokines. Under pathophysiological conditions, cardiac fibroblasts are involved in scar formation after myocardial infarction, cardiac fibrosis, and cardiac hypertrophy. HCF can be used to study such processes in vitro.
Catalog No. 50-306-008 Supplier Promocell GmbH Supplier No. C12375
Add to Cart
Add to Cart
Primary Human Cardiac Fibroblasts (HCF) are isolated from the ventricles of the adult heart. They play a central role in the maintenance of the extracellular matrix in the normal heart and the synthesis of growth factors and cytokines. Under pathophysiological conditions, cardiac fibroblasts are involved in scar formation after myocardial infarction, cardiac fibrosis, and cardiac hypertrophy. HCF can be used to study such processes in vitro.