Learn More
Aviva Systems Biology Corp HCFC1 Antibody - N-terminal region (ARP38600_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38600_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HCFC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HCFC1. Alternate names: CFF, HCF1, HFC1, VCAF, HCF-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53110