Learn More
Aviva Systems Biology Corp HCFC1 Antibody - middle region (ARP38601_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38601_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HCFC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HCFC1. Alternate names: CFF, HCF-1, HCF1, HFC1, MGC70925, VCAFApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56722