Learn More
Aviva Systems Biology Corp HCFC1R1 Antibody - middle region (ARP56149_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56149_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HCFC1R1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HCFC1R1. Alternate names: FLJ20568, HPIP, MGC70711, MGC99622ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-61740