Learn More
Aviva Systems Biology Corp hCG_1646157 Antibody - middle region (ARP50952_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50952_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against hCG_1646157. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human hCG_1646157. Alternate names: LOC440122, hCG_1646157ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53644