Learn More
Aviva Systems Biology Corp hCG_1745121 Antibody - N-terminal region (ARP58634_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58634_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against hCG_1745121. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human hCG_1745121. Alternate names: LOC729920, Nip, hCG_1745121ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62382