Learn More
Aviva Systems Biology Corp hCG_2007354 Antibody - N-terminal region (ARP51074_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51074_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against hCG_2007354. It was validated on Western Blot. LOC647589,hypothetical protein LOC647589,hypothetical protein LOC647589,hypothetical protein LOC647589,hCG2007354The immunogen is a synthetic peptide directed towards the n terminal region of human hCG_2007354. Alternate names: ApplicationWB. Species reactivity: Dog, Horse, Human, Rabbit
Catalog No. 50-285-554