Learn More
Rockland Immunochemicals hCG Alpha chain Antibody 25µL
SDP
Supplier:  Rockland Immunochemicals 600101MR6S
Encompass
Anti-Human Chorionic Gonadotropin (hCG) Alpha Antibody has been tested in ELISA and WB.  Expect a band at ~14.9kDa for the alpha chain in western blot using appropriate tissues and lysates. ELISA reactivity expected against alpha hCG with an expected titer of ~1:100,000 and full hCG at ~1:50,000 dilution.
Catalog No. 50-202-4074