Learn More
ILEX LIFE SCIENCES LLC Ilex Life Sciences Human Chorionic Gonadotropin (hCG), Native Protein (Urine), Bioactive, 5 mg
SDP
Catalog No. 502119969
Encompass_Preferred
Our Human Chorionic Gonadotropin (hCG) is a native protein produced from a sterile preparation of placental glucoprotein urine of pregnant women and has a total Mw of 36.7 kDa. The hCG consists of 237 amino acids; 92 a.a. a-chain and 145 a.a. b-chain. Lyophilized with no additives. Free of HbsAg and antibodies to HIV and HCV. Biological Activity: Each lot is bioactivity tested and the IU/mg is stated on a lot-specific datasheet and vial label. Generally speaking, bioactivity ranges between 4000-6000 IU/mg. Application: Our hCG is commonly used in combination with our Pregnant Mare Serum Gonadotropin (PMSG, eCG) in superovulation protocols for production of genetically modified laboratory animals, such as GEMM by pronuclear microinjection (transgenic mouse), targeted mutations through CRISPR-Cas9 methods, and ES-cell-based gene targeting to produce knockout mice. Our hCG can be used in the generation of Xenopus laevis and X. tropicalis eggs or embryos for use in experimental models.
Catalog No. 50-211-9969 Supplier ILEX LIFE SCIENCES LLC Supplier No. A225005
Our Human Chorionic Gonadotropin (hCG) is a native protein produced from a sterile preparation of placental glucoprotein urine of pregnant women and has a total Mw of 36.7 kDa. The hCG consists of 237 amino acids; 92 a.a. a-chain and 145 a.a. b-chain. Lyophilized with no additives. Free of HbsAg and antibodies to HIV and HCV. Biological Activity: Each lot is bioactivity tested and the IU/mg is stated on a lot-specific datasheet and vial label. Generally speaking, bioactivity ranges between 4000-6000 IU/mg. Application: Our hCG is commonly used in combination with our Pregnant Mare Serum Gonadotropin (PMSG, eCG) in superovulation protocols for production of genetically modified laboratory animals, such as GEMM by pronuclear microinjection (transgenic mouse), targeted mutations through CRISPR-Cas9 methods, and ES-cell-based gene targeting to produce knockout mice. Our hCG can be used in the generation of Xenopus laevis and X. tropicalis eggs or embryos for use in experimental models.