Learn More
Crystal Chem Inc Human Chorionic Gonadotropin (HCG) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80958
Encompass
The HCG ELISA is a highly sensitive assay that is used to quantify levels of human HCG in serum. Highlights: Kit uses sample size of only 25 µL. High sensitivity of 0.7 IU/L. Dynamic range of 2.5 - 500 IU/L. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7864