Learn More
Drg International Inc Hcg Elisa Ruo
Supplier:  Drg International Inc EIA1469R
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of intact human chorionic gonadotropin (hCG) in serum or plasma. This product is for Research Use Only. The DRG HCG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal [mouse] antibody directed towards a unique antigenic site on an hCG molecule.  An aliquot of sample containing endogenous hCG is incubated in the coated well with enzyme conjugate, which is a monoclonal antibody directed against the alpha-chain of hCG conjugated with horseradish peroxidase.
Catalog No. 50-194-7536