Learn More
Drg International Inc DRG β-HCG ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA1911R
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of total human chorionic gonadotropin (hCG and b-hCG) in serum. This product is for Research Use Only. The DRG Beta-HCG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal antibody [mouse] directed towards a unique antigenic site on a Beta-HCG molecule.  An aliquot of sample containing endogenous Beta-HCG and/or HCG is incubated in the coated well with enzyme conjugate, which is an anti- Beta-HCG antibody conjugated with horseradish peroxidase. 
Catalog No. 50-187-1248