Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories hCG Serum Controls, Tri-Level

Catalog No. 23111300
Change view
Click to view available options
Quantity:
3 x 2.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-300 3 x 2.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-300 Supplier Germaine™ Laboratories Supplier No. 71003
Add to Cart
Add to Cart
This item is not returnable. View return policy

Germaine™ Laboratories hCG Serum Controls, Tri-Level are intended for use with qualitative hCG test procedures. For professional use

  • For use with qualitative hCG serum tests
  • Positive hCG at approx. 250mIU/mL w/ preservatives
  • Low Positive hCG at approx. 25mIU/mL w/ preservatives
  • Negative hCG at no detectable level of hCG with preservatives

Specifications

Content And Storage 2°C to 8°C
For Use With (Application) Serum
Quantity 3 x 2.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.