Learn More
Abbott™ hCG Urine Cassette
Catalog No. 23875556C Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
30 Tests
Catalog No. Quantity
23875556C 30 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23875556C Supplier Abbott™ Supplier No. 92210
Add to Cart
Add to Cart
This item is not returnable. View return policy
Superior performance for the qualitative detection of the pregnancy hormone, hCG (human chorionic gonadotropin), in urine. Single-step test provides positive results in as little as 3 minutes.
  • Sensitivity: 20mIU/mL
  • Highly sensitive assay with built-in control ensures accuracy
  • Convenient cassette format means no potentially messy dipsticks
Specifications
CPT Code 81025
Clia Complexity Waived
Detectable Analytes hCG
Sample Type Urine
Type hCG Urine Cassette Test
Quantity 30 Tests
Test Time 3 min.