Learn More
enQuire Bioreagents HCG-beta Anti-Human Monoclonal [Clone HCGb/54], Size: 100ug
Supplier:  enQuire Bioreagents 1082MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: HCGb/54. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: JAR or TT Cells. Placenta. Cellular Staining: Cytoplasmic, secreted. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant hCG beta protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 22kDa.Chromosome: 19q13.33. Gene Symbol: CGB. Gene ID: 1082. Ensemble: ENSG00000104827. Uniprot: NP_000728.
Catalog No. 50-188-4591