Learn More
Promocell GmbH PromoCell HCH-c 500,000 Human Chondrocytes, cryopreserved
Supplier:  Promocell GmbH C12710
Encompass
Primary Human Chondrocytes (HCH) are isolated from normal human articular cartilage from the knee and hip joints (lot specific source information is available on request). PromoCell HCH are routinely tested for their capacity to differentiate in 3D spheroids.The articular cartilage covers the joints between bones and contains no blood vessels, lymphatic vessels, or nerve fibers. It is composed of only one specialized cell type, the chondrocytes, which produce and maintain the extracellular matrix of cartilage, i.e. collagen (mostly type II) and proteoglycans (primarily aggrecan). Our HCH are now also available from HLA-typed donors.
Catalog No. 50-306-058