Learn More
Promocell GmbH PromoCell HCH Pellet from > 1 million Normal Human Chondrocytes
Catalog No. 50306154
Encompass
Cell pellet consisting of 1 million cells dissolved in 200µl RNAlater for subsequent RNA, DNA or protein analysis. Note: Cell pellets cannot be revived.Our Cell Pellets are stored in RNAlater – an aqueous protection solution that rapidly permeates the cells and stabilizes cellular DNA, RNA, and proteins and protects them from degradation. This makes our Cell Pellets very useful for a wide range of research applications like gene expression analysis, cell characterization studies, and protein characterization experiments.Each Cell Pellet is made from 1 million cells and is dissolved in 200 µl RNAlater. The Cell Pellets are produced from released cell lots and have the same characteristics, i.e. expression profile or cell type specific antigens. On request, viable cells from the same lot are available for cultivation.
Catalog No. 50-306-154 Supplier Promocell GmbH Supplier No. C14070
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Cell pellet consisting of 1 million cells dissolved in 200µl RNAlater for subsequent RNA, DNA or protein analysis. Note: Cell pellets cannot be revived.Our Cell Pellets are stored in RNAlater – an aqueous protection solution that rapidly permeates the cells and stabilizes cellular DNA, RNA, and proteins and protects them from degradation. This makes our Cell Pellets very useful for a wide range of research applications like gene expression analysis, cell characterization studies, and protein characterization experiments.Each Cell Pellet is made from 1 million cells and is dissolved in 200 µl RNAlater. The Cell Pellets are produced from released cell lots and have the same characteristics, i.e. expression profile or cell type specific antigens. On request, viable cells from the same lot are available for cultivation.