Learn More
Medchemexpress LLC HY-101283 5mg Medchemexpress, HCH6-1 CAS:1435265-06-7 Purity:>98%
SDP
Catalog No. 502029488
Encompass_Preferred
Medchemexpress, HY-101283 5mg HCH6-1 CAS:1435265-06-7 HCH6-1 is a competitive antagonist of Formyl peptide receptor 1 (FPR1), an emerging therapeutic target for the discovery of drugs to treat neutrophilic inflammatory diseases. HCH6-1 is a selective FPR1 inhibitor and has protective effects against acute lung injury (ALI). HCH6-1 inhibits superoxide anion generation, elastase release, and chemotaxis in human neutrophils activated by fMLF (an FPR1 agonist). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9488 Supplier Medchemexpress LLC Supplier No. HY1012835MG
Medchemexpress, HY-101283 5mg HCH6-1 CAS:1435265-06-7 HCH6-1 is a competitive antagonist of Formyl peptide receptor 1 (FPR1), an emerging therapeutic target for the discovery of drugs to treat neutrophilic inflammatory diseases. HCH6-1 is a selective FPR1 inhibitor and has protective effects against acute lung injury (ALI). HCH6-1 inhibits superoxide anion generation, elastase release, and chemotaxis in human neutrophils activated by fMLF (an FPR1 agonist). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.