Learn More
Eurofins Discoverx Hck, active, 10 µg. (14-577)
Supplier:  Eurofins Discoverx 14577
Encompass
N-terminal 6xHis-tagged recombinant, human Hck amino acids 230-497. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8995