Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Reagent 71, 0.05N Hydrochloric Acid in Methanol (1L)
SDP

Catalog No. NC9885118
Encompass_Preferred

Reagent 71, 0.05N Hydrochloric Acid in Methanol (1L)

Catalog No. NC9885118 Supplier Aqua Solutions Supplier No. DC875245
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent 71, 0.05N Hydrochloric Acid in Methanol (1L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.