Learn More
Enzo Life Sciences HCl, (0.1M) (100ml)
SDP
Catalog No. 502006806
Encompass_Preferred
Assay diluent and lysis reagent for cyclic nucleotides. Formulation: Liquid. 0.1M hydrochloric acid. Long Term Storage: Ambient.
Catalog No. 50-200-6806 Supplier Enzo Life Sciences Supplier No. ADI800082
Assay diluent and lysis reagent for cyclic nucleotides. Formulation: Liquid. 0.1M hydrochloric acid. Long Term Storage: Ambient.