Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions HCL 0.5N 500 ML
SDP

Catalog No. NC0423999
Encompass_Preferred

Hydrochloric Acid 0.5 Normal Aqueous (500ml)

Catalog No. NC0423999 Supplier Aqua Solutions Supplier No. 4190500ML
May include imposed supplier surcharges.
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrochloric Acid 0.5 Normal Aqueous (500ml)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.