Learn More
Sigma Aldrich Fine Chemicals Biosciences Hydrochloric acid | 7647-01-0 | MFCD00011324 | 100 mL
Catalog No. NC0909595
Encompass
Hydrochloric acid | Purity: ACS reagent, 37% | Mol Wt: 36.46 | 7647-01-0 | MFCD00011324 | 100 mL
Catalog No. NC0909595 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 258148100ML
Hydrochloric acid | Purity: ACS reagent, 37% | Mol Wt: 36.46 | 7647-01-0 | MFCD00011324 | 100 mL