Learn More
Cayman Chemical HCl asy Reagent20 m 1mL
SDP
Catalog No. 502896584
Encompass_Preferred
An ENPP1 and ENPP3 inhibitor (IC50s 0.59 and 0.62 UM respectively for the human enzymes)
Catalog No. 50-289-6584 Supplier Cayman Chemical Supplier No. 7000121 ML
Add to Cart
Add to Cart
An ENPP1 and ENPP3 inhibitor (IC50s 0.59 and 0.62 UM respectively for the human enzymes)