Learn More
Azzota Corp hcl lamp Ba 2in 51mm
Catalog No. 5000887055
Encompass_Preferred
2 (51mm) diameter Hollow Cathode Lamp for Perkin Elmer atomic absorption Barium-Ba
Catalog No. 50-008-87055 Supplier Azzota Corp Supplier No. SHC20BA
2 (51mm) diameter Hollow Cathode Lamp for Perkin Elmer atomic absorption Barium-Ba