Learn More
Azzota Corp hcl lamp Boron 1.5in
Catalog No. 5000887157
Encompass_Preferred
1 5 Boron-B Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm
Catalog No. 50-008-87157 Supplier Azzota Corp Supplier No. SHC15B
1 5 Boron-B Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm