Learn More
Azzota Corp hcl lamp Ce 2in 51mm
Catalog No. 5000887141
Encompass_Preferred
2 (51mm) diameter Hollow Cathode Lamp for Perkin Elmer atomic absorption Cerium (Ce)
Catalog No. 50-008-87141 Supplier Azzota Corp Supplier No. SHC20CE
2 (51mm) diameter Hollow Cathode Lamp for Perkin Elmer atomic absorption Cerium (Ce)