Learn More
Azzota Corp hcl lamp Cerium 1.5in
Catalog No. 5000887113
Encompass_Preferred
1 5 Cerium-Ce Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm
Catalog No. 50-008-87113 Supplier Azzota Corp Supplier No. SHC15CE
1 5 Cerium-Ce Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm