Learn More
Azzota Corp hcl lamp Dysprosium 1.5in
Catalog No. 5000887187
Encompass_Preferred
1 5 Dysprosium-DY Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm
Catalog No. 50-008-87187 Supplier Azzota Corp Supplier No. SHC15DY
1 5 Dysprosium-DY Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm