Learn More
Azzota Corp hcl lamp Eu 2in 51mm
Catalog No. 5000887142
Encompass_Preferred
2 (51mm) diameter Hollow Cathode Lamp for Perkin Elmer atomic absorption Europium (Eu) 4 pins coded
Catalog No. 50-008-87142 Supplier Azzota Corp Supplier No. SHC20EU
2 (51mm) diameter Hollow Cathode Lamp for Perkin Elmer atomic absorption Europium (Eu) 4 pins coded