Learn More
Azzota Corp hcl lamp Lanthanum 1.5in
Catalog No. 5000887161
Encompass_Preferred
1 5 Lanthanum (La) Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm
Catalog No. 50-008-87161 Supplier Azzota Corp Supplier No. SHC15LA
1 5 Lanthanum (La) Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm