Learn More
Azzota Corp hcl lamp Lutetium 1.5in
Catalog No. 5000887174
Encompass_Preferred
1 5 Lutetium (Lu) Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm
Catalog No. 50-008-87174 Supplier Azzota Corp Supplier No. SHC15LU
Add to Cart
Add to Cart
1 5 Lutetium (Lu) Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm