Learn More
Azzota Corp hcl lamp Palladium 1.5in
Catalog No. 5000887095
Encompass_Preferred
1 5 Palladium-Pd Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm
Catalog No. 50-008-87095 Supplier Azzota Corp Supplier No. SHC15PD
1 5 Palladium-Pd Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm