Learn More
Azzota Corp hcl lamp Rhenium 1.5in
Catalog No. 5000887140
Encompass_Preferred
1 5 Rhenium (Re) Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm
Catalog No. 50-008-87140 Supplier Azzota Corp Supplier No. SHC15RE
Add to Cart
Add to Cart
1 5 Rhenium (Re) Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm