Learn More
Azzota Corp hcl lamp Rubidium 1.5in
Catalog No. 5000887117
Encompass_Preferred
1 5 Rubidium (Rb) Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm
Catalog No. 50-008-87117 Supplier Azzota Corp Supplier No. SHC15RB
1 5 Rubidium (Rb) Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm