Learn More
Azzota Corp hcl lamp Selenium 1.5in
Catalog No. 5000887053
Encompass_Preferred
1 5 Selenium-Se Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm
Catalog No. 50-008-87053 Supplier Azzota Corp Supplier No. SHC15SE
Add to Cart
Add to Cart
1 5 Selenium-Se Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm