Learn More
Azzota Corp hcl lamp Silicon 1.5in
Catalog No. 5000887118
Encompass_Preferred
1 5 Silicon - Si Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm
Catalog No. 50-008-87118 Supplier Azzota Corp Supplier No. SHC15SI
1 5 Silicon - Si Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm