Learn More
Azzota Corp hcl lamp Tellurium 1.5in
Catalog No. 5000887114
Encompass_Preferred
1 5 Tellurium (Te) Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm
Catalog No. 50-008-87114 Supplier Azzota Corp Supplier No. SHC15TE
1 5 Tellurium (Te) Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm