Learn More
Azzota Corp hcl lamp Thallium 1.5in
Catalog No. 5000887191
Encompass_Preferred
1 5 Thallium - Tl Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm
Catalog No. 50-008-87191 Supplier Azzota Corp Supplier No. SHC15TL
1 5 Thallium - Tl Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm