Learn More
Azzota Corp hcl lamp Vanadium 1.5in
Catalog No. 5000887192
Encompass_Preferred
1 5 Vanadium - V Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm
Catalog No. 50-008-87192 Supplier Azzota Corp Supplier No. SHC15V
1 5 Vanadium - V Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm